missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylmethane, 96%

Catalog No. AAA1470614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470614 25 g
AAA1470603 1 g
AAA1470606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1470614 Supplier Thermo Scientific Chemicals Supplier No. A1470614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 630-76-2 | C25H20 | 320.435 g/mol

Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 630-76-2
Molecular Formula C25H20
Molecular Weight (g/mol) 320.435
MDL Number MFCD00014428
InChI Key PEQHIRFAKIASBK-UHFFFAOYSA-N
Synonym tetraphenylmethane, methane, tetraphenyl, tetraphenyl methane, benzene, 1,1',1,1'-methanetetrayltetrakis, triphenylmethyl benzene, benzene, 1,1',1,1'-methanethetrayltetrakis, acmc-20ajiz, pubchem10677
PubChem CID 12424
IUPAC Name tritylbenzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 280°C to 285°C
Boiling Point 431°C
Quantity 25 g
Beilstein 1913557
Solubility Information Insoluble in water.
Formula Weight 320.44
Percent Purity 96%
Chemical Name or Material Tetraphenylmethane
Hazard Category H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 211-144-0
RTECSNumber PB4050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only