accessibility menu, dialog, popup

UserName

CAS RN 6292-59-7

CH3H3CCH3SNH2OO

CAS RN 6292-59-7

IUPAC Name: 4-tert-butylbenzene-1-sulfonamide
Synonyms: 4-tert-butylbenzenesulfonamide
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: KYDZEZNYRFJCSA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results