missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonamide 98.0+%, TCI America™

Catalog No. B38375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38375G Supplier TCI America Supplier No. B38375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6292-59-7
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.295
MDL Number MFCD00068599
InChI Key KYDZEZNYRFJCSA-UHFFFAOYSA-N
PubChem CID 222872
IUPAC Name 4-tert-butylbenzenesulfonamide
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzenesulfonamide