CAS RN 629-78-7
Heptadecane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Heptadecane belongs to the n-alkane class of petroleum hydrocarbons.
n-Heptadecane, 99.5%
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.475 MDL Number: MFCD00009002 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N Synonym: n-heptadecane,heptadekan,unii-h7c0j39xum,hexadecane, methyl,h7c0j39xum,heptadecane, analytical standard,normal-heptadecane,heptadecane,dsstox_cid_27061,dsstox_rid_82078 PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC
n-Heptadecane, 99%
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.475 MDL Number: MFCD00009002 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N Synonym: n-heptadecane,heptadekan,unii-h7c0j39xum,hexadecane, methyl,h7c0j39xum,heptadecane, analytical standard,normal-heptadecane,heptadecane,dsstox_cid_27061,dsstox_rid_82078 PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC
n-Heptadecane, 99%
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.47 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N Synonym: n-heptadecane,heptadekan,unii-h7c0j39xum,hexadecane, methyl,h7c0j39xum,heptadecane, analytical standard,normal-heptadecane,heptadecane,dsstox_cid_27061,dsstox_rid_82078 PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC
Heptadecane 99.0+%, TCI America™
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.475 MDL Number: MFCD00009002 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N Synonym: n-heptadecane,heptadekan,unii-h7c0j39xum,hexadecane, methyl,h7c0j39xum,heptadecane, analytical standard,normal-heptadecane,heptadecane,dsstox_cid_27061,dsstox_rid_82078 PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC
Heptadecane 99.5+%, TCI America™
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.475 MDL Number: MFCD00009002 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N Synonym: n-heptadecane,heptadekan,unii-h7c0j39xum,hexadecane, methyl,h7c0j39xum,heptadecane, analytical standard,normal-heptadecane,heptadecane,dsstox_cid_27061,dsstox_rid_82078 PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC
n-Heptadecane, EMD-Millipore
CAS: 629-78-7 Molecular Formula: C17H36 Molecular Weight (g/mol): 240.475 InChI Key: NDJKXXJCMXVBJW-UHFFFAOYSA-N PubChem CID: 12398 ChEBI: CHEBI:16148 IUPAC Name: heptadecane SMILES: CCCCCCCCCCCCCCCCC