missing translation for 'onlineSavingsMsg'
Learn More

Heptadecane 99.5+%, TCI America™

Catalog No. S02895ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02895ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02895ML Supplier TCI America Supplier No. S02895ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Color White
Boiling Point 302°C
Quantity 5 mL
Formula Weight 240.48
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Heptadecane [Standard Material for GC]
EINECSNumber (2)-0010
RTECSNumber MI3550000
TSCA Yes