missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, 99%

Catalog No. AC156245000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156245000 500 mL Glass bottle
AC156240250 25 mL Glass bottle
AC156241000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156245000 Supplier Thermo Scientific Chemicals Supplier No. 156245000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-78-7 | C17H36 | 240.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.47
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Melting Point 22.0°C to 24.0°C
Color Colorless
Density 0.7780g/mL
Boiling Point 302.0°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370
Linear Formula CH3(CH2)15CH3
Quantity 500 mL
Beilstein 01,173
Specific Gravity 0.778
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 240.47
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material n-Heptadecane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 211-108-4

RUO – Research Use Only