missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, EMD-Millipore

Catalog No. M1096040005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096040005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096040005 Supplier Technidata Supplier No. 1.09604.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrocarbons C9 – C19

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Packaging Glass ampoule
Vapor Pressure < - 0.1 hPa (20°C)
Viscosity <7mm2/sec., kinematic (40°C)
Density Density: 0.78g/cm3 (20°)
Assay Percent Range ≥99.3%
Linear Formula CH3(CH2)15CH3
Quantity 5 mL
Formula Weight 240.47
Chemical Name or Material n-Heptadecane
Recommended Storage Store without restrictions