CAS RN 629-73-2
1-Hexadecene, 90+%
CAS: 629-73-2 Molecular Formula: C16H32 Molecular Weight (g/mol): 224.432 MDL Number: MFCD00008991 InChI Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N Synonym: 1-hexadecene,cetene,1-cetene,hexadecylene-1,hexadecene,1-n-hexadecene,cetylene,n-hexadec-1-ene,alpha-hexadecene,alpha-hexadecylene PubChem CID: 12395 ChEBI: CHEBI:77507 IUPAC Name: hexadec-1-ene SMILES: CCCCCCCCCCCCCCC=C
1-Hexadecene, 92%, tech.
CAS: 629-73-2 Molecular Formula: C16H32 Molecular Weight (g/mol): 224.43 MDL Number: MFCD00008991 InChI Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N Synonym: 1-hexadecene,cetene,1-cetene,hexadecylene-1,hexadecene,1-n-hexadecene,cetylene,n-hexadec-1-ene,alpha-hexadecene,alpha-hexadecylene PubChem CID: 12395 ChEBI: CHEBI:77507 IUPAC Name: hexadec-1-ene SMILES: CCCCCCCCCCCCCCC=C
1-Hexadecene 99.0+%, TCI America™
CAS: 629-73-2 Molecular Formula: C16H32 Molecular Weight (g/mol): 224.432 MDL Number: MFCD00008991 InChI Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N Synonym: 1-hexadecene,cetene,1-cetene,hexadecylene-1,hexadecene,1-n-hexadecene,cetylene,n-hexadec-1-ene,alpha-hexadecene,alpha-hexadecylene PubChem CID: 12395 ChEBI: CHEBI:77507 IUPAC Name: hexadec-1-ene SMILES: CCCCCCCCCCCCCCC=C
1-Hexadecene 90.0+%, TCI America™
CAS: 629-73-2 Molecular Formula: C16H32 Molecular Weight (g/mol): 224.432 MDL Number: MFCD00008991 InChI Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N Synonym: 1-hexadecene,cetene,1-cetene,hexadecylene-1,hexadecene,1-n-hexadecene,cetylene,n-hexadec-1-ene,alpha-hexadecene,alpha-hexadecylene PubChem CID: 12395 ChEBI: CHEBI:77507 IUPAC Name: hexadec-1-ene SMILES: CCCCCCCCCCCCCCC=C
1-Hexadecene 99.5+%, TCI America™
CAS: 629-73-2 Molecular Formula: C16H32 Molecular Weight (g/mol): 224.432 MDL Number: MFCD00008991 InChI Key: GQEZCXVZFLOKMC-UHFFFAOYSA-N Synonym: 1-hexadecene,cetene,1-cetene,hexadecylene-1,hexadecene,1-n-hexadecene,cetylene,n-hexadec-1-ene,alpha-hexadecene,alpha-hexadecylene PubChem CID: 12395 ChEBI: CHEBI:77507 IUPAC Name: hexadec-1-ene SMILES: CCCCCCCCCCCCCCC=C