missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene, 90+%

Catalog No. AAL02637AU
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL02637AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL02637AU Supplier Thermo Scientific Chemicals Supplier No. L02637AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-73-2 | C16H32 | 224.432 g/mol

1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

Solubility
Miscible with ether, petroleum ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Melting Point 4°C
Density 0.781
Boiling Point 273°C to 274°C
Flash Point 132°C (269°F)
Refractive Index 1.442
Quantity 1000 mL
Beilstein 1753401
Solubility Information Miscible with ether,petroleum ether and alcohol. Immiscible with water.
Formula Weight 224.43
Percent Purity 94%
Chemical Name or Material 1-Hexadecene
Show More Show Less
Hazard Category H304
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 211-105-8
RTECSNumber MM0479000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only