missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014952
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014952 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014952 Supplier Merck Emd Millipore Supplier No. 522765ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Petroleum

Specifications

CAS 629-73-2
Ignition Point 464°F
Vapor Pressure <0.2 mmHg (20°C)
Vapor Density >0.8 (vs. air)
Density 0.783 g/mL (at 25°C)
Flash Point 132°C
Physical Form Neat
Linear Formula CH3(CH2)13 CH=CH2
MDL Number MFCD00008991
Melting Point 3°C to 5°C
Boiling Point 274°C
Synonym Cetene
Type Analytical Standard
Quantity 5 mL
Formula Weight 224.43
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less