missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene 99.5+%, TCI America™

Catalog No. S03465ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03465ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03465ML Supplier TCI America Supplier No. S03465ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 285°C
Quantity 5 mL
Formula Weight 224.43
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecene [Standard Material for GC]
EINECSNumber (2)-0027
RTECSNumber MM0479000
TSCA Yes