accessibility menu, dialog, popup

UserName

H3CNNOHN(R)OOOHOO

CAS RN 62893-24-7

IUPAC Name: (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetate
Synonyms: (2R)-2-(4-ethyl-2,3-dioxopiperazine-1-carbonylamino)-2-(4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 334.31
Molecular Formula: C15H16N3O6
InChi Key: IPARGUVYMOMVNU-LLVKDONJSA-M
SMILES: CCN1CCN(C(=O)N[C@@H](C([O-])=O)C2=CC=C(O)C=C2)C(=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results