missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic Acid 98.0+%, TCI America™

Catalog No. E09945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09945G 5 g
E09941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09945G Supplier TCI America Supplier No. E09945G
Only null left

Chemical Identifiers

CAS 62893-24-7
Molecular Formula C15H17N3O6
Molecular Weight (g/mol) 335.316
MDL Number MFCD00792472
InChI Key IPARGUVYMOMVNU-LLVKDONJSA-N
Synonym (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic Acid
PubChem CID 2733657
IUPAC Name (2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetic acid
SMILES CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)O

Specifications

Melting Point 203°C
Color White
Quantity 5 g
Formula Weight 335.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic Acid