accessibility menu, dialog, popup

UserName

CAS RN 628-87-5

IUPAC Name: 2-[(cyanomethyl)amino]acetonitrile
Synonyms: acetonitrile, 2,2'-iminobis-
Molecular Weight (g/mol): 95.11
Molecular Formula: C4H5N3
InChi Key: BSRDNMMLQYNQQD-UHFFFAOYSA-N
SMILES: N#CCNCC#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)