missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetonitrile 98.0+%, TCI America™

Catalog No. D0434500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0434500G 500 g
D043425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0434500G Supplier TCI America Supplier No. D0434500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-87-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00001887
InChI Key BSRDNMMLQYNQQD-UHFFFAOYSA-N
Synonym iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
PubChem CID 69413
ChEBI CHEBI:71189
IUPAC Name 2-[(cyanomethyl)amino]acetonitrile
SMILES N#CCNCC#N

Specifications

Melting Point 78°C
Color White
Quantity 500 g
Formula Weight 95.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Iminodiacetonitrile
RTECSNumber AM0650000
TSCA Yes