accessibility menu, dialog, popup

UserName

CAS RN 628-21-7

IUPAC Name: 1,4-diiodobutane
Synonyms: butane, 1,4-diiodo-
Molecular Weight (g/mol): 309.92
Molecular Formula: C4H8I2
InChi Key: ROUYUBHVBIKMQO-UHFFFAOYSA-N
SMILES: ICCCCI
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)