accessibility menu, dialog, popup

UserName

H3CI

CAS RN 628-17-1

IUPAC Name: 1-iodopentane
Synonyms: 1-iodopentane
Molecular Weight (g/mol): 198.05
Molecular Formula: C5H11I
InChi Key: BLXSFCHWMBESKV-UHFFFAOYSA-N
SMILES: CCCCCI
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)