missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I006625ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006625ML 25 mL
I0066250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I006625ML Supplier TCI America Supplier No. I006625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-17-1
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.047
MDL Number MFCD00001100
InChI Key BLXSFCHWMBESKV-UHFFFAOYSA-N
Synonym n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
PubChem CID 12335
IUPAC Name 1-iodopentane
SMILES CCCCCI

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
UN Number 1993
Formula Weight 198.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodopentane (stabilized with Copper chip)
RTECSNumber SA2975600
TSCA Yes