accessibility menu, dialog, popup

UserName

CAS RN 62669-66-3

IUPAC Name: 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-2,7-dimethyl-10λ⁴-xanthen-10-ylium perchlorate
Synonyms: 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-2,7-dimethyl-10λ⁴-xanthen-10-ylium perchlorate
Molecular Weight (g/mol): 571.07
Molecular Formula: C30H35ClN2O7
InChi Key: WLXLQXVNKTVEJE-UHFFFAOYSA-N
SMILES: [O-][Cl](=O)(=O)=O.CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results