Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine 19 Perchlorate 98,0+%, TCI America™

Catalog No. R01915G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
R01915G 5 g
R01911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01915G Supplier TCI America Supplier No. R01915G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62669-66-3
Formule moléculaire C26H27ClN2O7
Poids moléculaire (g/mol) 514.959
Numéro MDL MFCD02684062
Clé InChI WRJTXSZPMAXPRF-UHFFFAOYSA-N
PubChem CID 45157597
Nom de l’IUPAC acide benzoïque 2-[3-(éthylamino)-6-éthylimino-2,7-diméthylxanthen-9-yl; Acide perchlorique
SOURIRES CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)O)C)C.OCl(=O)(=O)=O

Specifications

Nom ou substance chimique Rhodamine 19 Perchlorate
Quantité 5 g
Numéro UN 1479
Poids de formule 514.96
Pourcentage de pureté ≥98,0% (HPLC,N)
Forme physique Poudre cristalline