CAS RN 62625-21-2
Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue sodium salt, 0.04% w/v aq. soln.
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thermo Scientific Chemicals Thymol Blue sodium salt, ACS reagent
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thermo Scientific Chemicals Thymol Blue sodium salt
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M Synonym: Thymol Blue, water soluble PubChem CID: 23692293 IUPAC Name: sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue, 0.4% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue, Certified, LabChem™
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue Sodium Salt, ACS Grade, LabChem™
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue, 0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue, Certified, LabChem™
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue Indicator, 0.4% (w/v), Ricca Chemical
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): Mixture MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue Sodium Salt, ACS Reagent Grade, Ricca Chemical
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue Indicator, 0.04% (w/v), Ricca Chemical
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): Mixture MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Thymol Blue Sodium Salt, TCI America™
CAS: 62625-21-2 Molecular Formula: C27H29NaO5S Molecular Weight (g/mol): 488.57 MDL Number: MFCD00151093 InChI Key: OCMIKNBSIRSUPI-FRWNMSGJSA-M Synonym: Sodium Thymol Blue PubChem CID: 23692293 IUPAC Name: sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate SMILES: [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O