missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue, 0.4% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue, Certified, LabChem™

Catalog No. LC260407
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC260407 125 mL Poly Bottle
LC260401 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC260407 Supplier LabChem Supplier No. LC260407

Chemical Identifiers

CAS 62625-21-2 , 7732-18-5
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

CAS Max % 99.6, 0.4
Linear Formula C27H29NaO5S
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 488.58
Grade Certified
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Blue/Green
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Thymol Blue, 0.4% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature