missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue sodium salt, 0.04% w/v aq. soln.

Catalog No. AA38692AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38692AP 500 mL
AA38692AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38692AP Supplier Thermo Scientific Chemicals Supplier No. 038692AP

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 488.58
Odor Odorless
Density 0.979 g/mL
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Thymol Blue sodium salt
EINECSNumber 200-973-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only