accessibility menu, dialog, popup

UserName

H3CCH3HS

CAS RN 6262-87-9

IUPAC Name: 2-(propan-2-yl)benzene-1-thiol
Synonyms: 2-isopropylbenzenethiol
Molecular Weight (g/mol): 152.26
Molecular Formula: C9H12S
InChi Key: QEDRUXIMTJVXFL-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results