accessibility menu, dialog, popup

UserName

CAS RN 626-51-7

CH3HOOOHO

CAS RN 626-51-7

IUPAC Name: 3-methylpentanedioic acid
Synonyms: methylglutaric acid methylglutarate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: XJMMNTGIMDZPMU-UHFFFAOYSA-N
SMILES: CC(CC(O)=O)CC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)