accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 622-98-0

IUPAC Name: 1-chloro-4-ethylbenzene
Synonyms: benzene, 1-chloro-4-ethyl-
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: GPOFSFLJOIAMSA-UHFFFAOYSA-N
SMILES: CCC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)