missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-ethylbenzene 98.0+%, TCI America™

Catalog No. C30915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30915G 5 g
C309125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30915G Supplier TCI America Supplier No. C30915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-98-0
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00040855
InChI Key GPOFSFLJOIAMSA-UHFFFAOYSA-N
Synonym benzene, 1-chloro-4-ethyl, p-chloroethylbenzene, 4-chloroethylbenzene, 4-ethylchlorobenzene, acmc-20aovw, p-chlorophenyl ethane, 4-chloro ethylbenzene, 1-ethyl-4-chlorobenzene, 4-chloro-1-ethylbenzene, 1-chloro-4-ethyl benzene
PubChem CID 69330
IUPAC Name 1-chloro-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 184°C
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Chloro-4-ethylbenzene