accessibility menu, dialog, popup

UserName

CH3CH2

CAS RN 622-97-9

IUPAC Name: 1-ethenyl-4-methylbenzene
Synonyms: 4-vinyltoluene P-methylstyrene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: JLBJTVDPSNHSKJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C)C=C1
Boiling Point 
  • (4)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)