missing translation for 'onlineSavingsMsg'
Learn More

4-Methylstyrene, 98%, stab. with 0.1% 3,5-di-tert-butylcatechol

Catalog No. AAL0148018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0148018 50 g
AAL0148009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0148018 Supplier Thermo Scientific Chemicals Supplier No. L0148018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 622-97-9 | C9H10 | 118.18 g/mol

4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

Solubility
Miscible with benzene. slightly miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 622-97-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008621
InChI Key JLBJTVDPSNHSKJ-UHFFFAOYSA-N
Synonym 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
PubChem CID 12161
IUPAC Name 1-ethenyl-4-methylbenzene
SMILES CC1=CC=C(C=C)C=C1

Specifications

Melting Point -34°C
Density 0.896
Boiling Point 171°C to 175°C
Flash Point 45°C (113°F)
Refractive Index 1.543
Quantity 50 g
UN Number UN2618
Beilstein 1209317
Solubility Information Miscible with benzene. slightly miscible with water.
Formula Weight 118.18
Percent Purity ≥98%
Chemical Name or Material 4-Methylstyrene, stabilized with 0.1% 3,5-di-tert-butylcatechol
Show More Show Less
Hazard Category H226-H304-H315-H319-H335-H340
Hazard Statement GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED
EINECSNumber 210-762-8
RTECSNumber WL5076000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only