accessibility menu, dialog, popup

UserName

OHHO

CAS RN 620-92-8

IUPAC Name: 4-[(4-hydroxyphenyl)methyl]phenol
Synonyms: bis(4-hydroxyphenyl)methane HDM 4,4'-dihydroxydiphenylmethane 4,4'-methylene diphenol p,p'-bis(hydroxyphenyl)methane
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: PXKLMJQFEQBVLD-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)