missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxydiphenylmethane, 98%

Catalog No. AAA1141706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1141706 5 g
AAA1141703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1141706 Supplier Thermo Scientific Chemicals Supplier No. A1141706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 620-92-8 | C13H12O2 | 200.237 g/mol

4,4'-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

Solubility
It is soluble in ethanol, ether, chloroform, alkali; slightly soluble in DMSO, insoluble in carbon disulfide and water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 620-92-8
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002385
InChI Key PXKLMJQFEQBVLD-UHFFFAOYSA-N
Synonym 4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol
PubChem CID 12111
ChEBI CHEBI:34575
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)O)O

Specifications

Melting Point 159°C to 163°C
Quantity 5 g
Beilstein 2049425
Solubility Information It is soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO,insoluble in carbon disulfide and water.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 4,4'-Dihydroxydiphenylmethane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-658-2
RTECSNumber SL9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only