missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol F, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014918
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014918 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014918 Supplier Merck Emd Millipore Supplier No. 51453100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 620-92-8
Physical Form Neat
Linear Formula CH2(C6H4 OH)2
MDL Number MFCD00002385
Melting Point 162°C to 164°C
Synonym Bis(4-hydroxyphenyl)methane; 4,4′-Methylenediphenol; Bisphenol F
Type Analytical Standard
Quantity 100 mg
Formula Weight 200.23
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less