accessibility menu, dialog, popup

UserName

CAS RN 61857-83-8

IUPAC Name: 2,2-dimethyl-5-(thiophen-3-yl)-1,3-dioxane-4,6-dione
Synonyms: 2,2-dimethyl-5-(thiophen-3-yl)-1,3-dioxane-4,6-dione
Molecular Weight (g/mol): 226.25
Molecular Formula: C10H10O4S
InChi Key: HAGWJJFKVFSZLP-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)C(C2=CSC=C2)C(=O)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results