missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-5-(3-thienyl)-1,3-dioxane-4,6-dione 98.0+%, TCI America™

Catalog No. D375925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D375925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D375925G Supplier TCI America Supplier No. D375925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61857-83-8
Molecular Formula C10H10O4S
Molecular Weight (g/mol) 226.246
MDL Number MFCD08458279
InChI Key HAGWJJFKVFSZLP-UHFFFAOYSA-N
Synonym 5-(Thienyl)meldrum′Cs Acid
PubChem CID 370301
IUPAC Name 2,2-dimethyl-5-thiophen-3-yl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)C(C(=O)O1)C2=CSC=C2)C

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Formula Weight 226.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethyl-5-(3-thienyl)-1,3-dioxane-4,6-dione