accessibility menu, dialog, popup

UserName

H3CNNH2

CAS RN 618-40-6

IUPAC Name: 1-methyl-1-phenylhydrazine
Synonyms: hydrazine, 1-methyl-1-phenyl-
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: MWOODERJGVWYJE-UHFFFAOYSA-N
SMILES: CN(N)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)