accessibility menu, dialog, popup

UserName

CAS RN 617-54-9

CH3OOH3COOH3C(E)

CAS RN 617-54-9

IUPAC Name: 1,4-dimethyl (2E)-2-methylbut-2-enedioate
Synonyms: citraconate de dimethyle
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: WQEXBUQDXKPVHR-SNAWJCMRSA-N
SMILES: COC(=O)\C=C(/C)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results