missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Citraconate 99.0+%, TCI America™

Catalog No. C03645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03645G 5 g
C036425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03645G Supplier TCI America Supplier No. C03645G
Only null left

Chemical Identifiers

CAS 617-54-9
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00059306
InChI Key WQEXBUQDXKPVHR-SNAWJCMRSA-N
Synonym Citraconic Acid Dimethyl Ester
PubChem CID 5355715
IUPAC Name 1,4-dimethyl (2E)-2-methylbut-2-enedioate
SMILES COC(=O)\C=C(/C)C(=O)OC

Specifications

Color Yellow
Boiling Point 211°C
Quantity 5 g
Formula Weight 158.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Citraconate
RTECSNumber GE6700000
TSCA Yes