accessibility menu, dialog, popup

UserName

CAS RN 615-76-9

IUPAC Name: 6-methyl-3-sulfanylidene-2,3,4,5-tetrahydro-1,2,4-triazin-5-one
Synonyms: 2-thio-6-azathymine
Molecular Weight (g/mol): 143.16
Molecular Formula: C4H5N3OS
InChi Key: NKOPQOSBROLOFP-UHFFFAOYSA-N
SMILES: CC1=NNC(=S)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results