accessibility menu, dialog, popup

UserName

CH3NH2Cl

CAS RN 615-65-6

IUPAC Name: 2-chloro-4-methylaniline
Synonyms: 2-chloro-4-methylaniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: XGYLSRFSXKAYCR-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)