accessibility menu, dialog, popup

UserName

NH2I

CAS RN 615-43-0

IUPAC Name: 2-iodoaniline
Synonyms: 2-iodoaniline
Molecular Weight (g/mol): 219.03
Molecular Formula: C6H6IN
InChi Key: UBPDKIDWEADHPP-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1I
Percent Purity 
  • (4)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)