accessibility menu, dialog, popup

UserName

CAS RN 615-28-1

IUPAC Name: dihydrogen benzene-1,2-diamine dichloride
Synonyms: dihydrogen o-phenylenediamine dichloride
Molecular Weight (g/mol): 181.06
Molecular Formula: C6H10Cl2N2
InChi Key: RIIWUGSYXOBDMC-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)