missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine dihydrochloride, 98+%

Catalog No. AAJ6035422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6035422 100 g
AAJ6035414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6035422 Supplier Thermo Scientific Chemicals Supplier No. J6035422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A peroxidase substrate for ELISA procedures | CAS: 615-28-1 | C6H10Cl2N2 | 181.06 g/mol

o-Phenylenediamine is a peroxidase substrate suitable for use in ELISA procedures. The substrate produces a soluble end product that is orange-brown in color and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3 N HCl or 3 M H2SO4 and read at 492 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Phenylenediamine is a peroxidase substrate suitable for use in ELISA procedures. The substrate produces a soluble end product that is orange-brown in color and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3 N HCl or 3 M H2SO4 and read at 492 nm.

Solubility
Slightly soluble in water; freely soluble in alcohol, chloroform or ether

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

Chemical Identifiers

CAS 615-28-1
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00012966
InChI Key RIIWUGSYXOBDMC-UHFFFAOYSA-N
Synonym 1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
PubChem CID 11990
IUPAC Name benzene-1,2-diamine;dihydrochloride
SMILES C1=CC=C(C(=C1)N)N.Cl.Cl

Specifications

Quantity 100 g
UN Number UN2811
Beilstein 3912045
Solubility Information Slightly soluble in water; freely soluble in alcohol,chloroform or ether
Formula Weight 181.07
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material o-Phenylenediamine dihydrochloride
Hazard Category H301-H312+H332-H317-H319-H341-H351
Hazard Statement GHS H Statement
H301-H341-H351-H312-H332-H319-H317
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-418-7
RTECSNumber ST0175000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only