accessibility menu, dialog, popup

UserName

H3CCH3OOO

CAS RN 615-12-3

IUPAC Name: 3-methylbutyl furan-2-carboxylate
Synonyms: 3-methylbutyl furan-2-carboxylate
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: FPBZWZKAUPNMMV-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C1=CC=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results