missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl 2-Furancarboxylate (contains 2-Methylbutyl 2-Furancarboxylate) 80.0+%, TCI America™

Catalog No. F0156500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0156500ML 500 mL
F015625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0156500ML Supplier TCI America Supplier No. F0156500ML
Only null left

Chemical Identifiers

CAS 615-12-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00059015
InChI Key FPBZWZKAUPNMMV-UHFFFAOYSA-N
Synonym 2-Furancarboxylic Acid Isoamyl Ester, Isoamyl 2-Furoate, 2-Furoic Acid Isoamyl Ester
PubChem CID 69203
IUPAC Name 3-methylbutyl furan-2-carboxylate
SMILES CC(C)CCOC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 232°C
Quantity 500 mL
Formula Weight 182.22
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl 2-Furancarboxylate (contains 2-Methylbutyl 2-Furancarboxylate)
EINECSNumber (5)-0046
TSCA No