accessibility menu, dialog, popup

UserName

CAS RN 614-65-3

OHNNH(E)

CAS RN 614-65-3

IUPAC Name: (6E)-6-[(2-phenylhydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-[(2-phenylhydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: AWSLIWUMMSIZFW-ZHACJKMWSA-N
SMILES: O=C1C=CC=C\C1=C/NNC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results