missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzaldehyde Phenylhydrazone 98.0+%, TCI America™

Catalog No. H14785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14785G 5 g
H147825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14785G Supplier TCI America Supplier No. H14785G
Only null left

Chemical Identifiers

CAS 614-65-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00136159
InChI Key AWSLIWUMMSIZFW-ZHACJKMWSA-N
Synonym Salicylaldehyde Phenylhydrazone
PubChem CID 10813780
IUPAC Name (6E)-6-[(2-phenylhydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C/NNC1=CC=CC=C1

Specifications

Melting Point 144°C
Boiling Point 234°C
Quantity 5 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzaldehyde Phenylhydrazone