CAS RN 61281-38-7
CAS RN 61281-38-7
IUPAC Name:
3,4,5,14,15,16-hexamethoxy-9,10-dimethyltricyclo[10.4.0.0²,⁷]hexadeca-1(12),2,4,6,13,15-hexaene
Synonyms:
3,4,5,14,15,16-hexamethoxy-9,10-dimethyltricyclo[10.4.0.0²,⁷]hexadeca-1(12),2,4,6,13,15-hexaene
Molecular Weight (g/mol):
416.51
Molecular Formula:
C24H32O6
InChi Key:
JEJFTTRHGBKKEI-UHFFFAOYNA-N
SMILES:
COC1=CC2=C(C(OC)=C1OC)C1=C(OC)C(OC)=C(OC)C=C1CC(C)C(C)C2
1
–
2
of
2
results
2
Schisandrin A, Thermo Scientific Chemicals
CAS: 61281-38-7 Molecular Weight (g/mol): 416.51