missing translation for 'onlineSavingsMsg'
Learn More

Schisandrin A, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014887
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014887 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014887 Supplier Merck Emd Millipore Supplier No. 9013950MG
Only null left

Specifications

CAS 61281-38-7
Linear Formula C24H32O6
MDL Number MFCD09026934
Synonym Deoxyschisandrin; Deoxyschizandrin; Schizandrin A
Type Analytical Standard
Quantity 50 mg
Formula Weight 416.51
Percent Purity ≥98% (HPLC)
Grade Analytical Standard