accessibility menu, dialog, popup

UserName

CAS RN 6112-76-1

IUPAC Name: 6,7-dihydro-3H-purine-6-thione hydrate
Synonyms: mercaptopurine hydrate
Molecular Weight (g/mol): 170.19
Molecular Formula: C5H6N4OS
InChi Key: WFFQYWAAEWLHJC-UHFFFAOYSA-N
SMILES: O.S=C1N=CNC2=C1NC=N2
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)