missing translation for 'onlineSavingsMsg'
Learn More

Mercaptopurine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018646
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018646 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018646 Supplier Merck Emd Millipore Supplier No. PHR16111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Mercaptopurine is an anticancer and an immunosuppressive drug employed for the management of acute lymphocytic leukemia (ALL), chronic myeloid leukemia (CML), Crohn′s disease, etc. Its mode of action involves interference in the nucleic acid synthesis by the inhibition of purine metabolism.

Specifications

CAS 6112-76-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C5H4N4 S·H2O
MDL Number MFCD03854445
Synonym 6-Mercaptopurine monohydrate; 6-Purinethiol; 6-Thiohypoxanthine
Type Certified Reference Material
Quantity 1 g
Formula Weight 170.19
Grade Certified Reference Material